missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene oxide, 98%, Thermo Scientific Chemicals

Product Code. 10707392
25 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-30-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00066354
InChI Key GELKGHVAFRCJNA-UHFFFAOYSA-N
Synonym isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxideShow MoreShow Less
PubChem CID 11208
IUPAC Name 2,2-dimethyloxirane
SMILES CC1(CO1)C

Specifications

Color Colorless
Density 0.8080g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3710 to 1.3730
Quantity 25 mL
Beilstein 17, V,1, 58
Specific Gravity 0.808
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether
Viscosity 0.513 mm2/s (20°C)
Formula Weight 72.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutylene oxide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutylene oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutylene oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.