Learn More
Isobutyllithium, 1.6M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
£100.40 - £351.00
Chemical Identifiers
CAS | 920-36-5, 64742-49-0 |
---|---|
Molecular Formula | C4H9Li |
Molecular Weight (g/mol) | 64.056 |
MDL Number | MFCD01863085 |
InChI Key | CCZVEWRRAVASGL-UHFFFAOYSA-N |
Synonym | isobutyllithium, isobutyllithium solution, 2-methylpropyl lithium, lithium 2-methanidylpropane, lithium 2-methyl-1-propanide, i-butyllithium, isobutyl lithium, i-buli, isobutyllithiumsolution, iso-butyllithium 0.7m in hexanes |
PubChem CID | 2734901 |
IUPAC Name | lithium;2-methanidylpropane |
SMILES | [Li+].CC(C)[CH2-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10026182
|
Thermo Scientific Acros
377591000 |
100 mL | AcroSeal™ Glass bottle |
£100.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10402493
|
Thermo Scientific Acros
377598000 |
800 mL | AcroSeal™ Glass Bottle |
£351.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
920-36-5, 64742-49-0 | |
64.056 | |
CCZVEWRRAVASGL-UHFFFAOYSA-N | |
2734901 | |
[Li+].CC(C)[CH2-] |
C4H9Li | |
MFCD01863085 | |
isobutyllithium, isobutyllithium solution, 2-methylpropyl lithium, lithium 2-methanidylpropane, lithium 2-methyl-1-propanide, i-butyllithium, isobutyl lithium, i-buli, isobutyllithiumsolution, iso-butyllithium 0.7m in hexanes | |
lithium;2-methanidylpropane |
Specifications
Yellow | |
C4H9Li | |
MFCD01863085 | |
isobutyllithium, isobutyllithium solution, 2-methylpropyl lithium, lithium 2-methanidylpropane, lithium 2-methyl-1-propanide, i-butyllithium, isobutyl lithium, i-buli, isobutyllithiumsolution, iso-butyllithium 0.7m in hexanes | |
CCZVEWRRAVASGL-UHFFFAOYSA-N | |
lithium;2-methanidylpropane | |
2734901 | |
15 to 17% (as iso-BuLi) | |
Liquid |
920-36-5, 64742-49-0 | |
(CH3)2CHCH2Li | |
0.7000g/mL | |
Solubility in water: vigorous reaction. | |
[Li+].CC(C)[CH2-] | |
64.056 | |
64.06 | |
0.7 | |
Isobutyllithium, 1.6M Solution in Heptane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment.
Danger
- Isobutyllithium, 1.6M solution in heptane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Pyrophoric liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only