Learn More
Isobutyltriethoxysilane, 95%, Thermo Scientific™
£70.70 - £230.00
Chemical Identifiers
CAS | 17980-47-1 |
---|---|
Molecular Formula | C10H24O3Si |
Molecular Weight (g/mol) | 220.384 |
MDL Number | MFCD00069084 |
InChI Key | ALVYUZIFSCKIFP-UHFFFAOYSA-N |
Synonym | triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilaneShow More |
PubChem CID | 87391 |
IUPAC Name | triethoxy(2-methylpropyl)silane |
SMILES | CCO[Si](CC(C)C)(OCC)OCC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used in a wide range of applications including silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent, silane cross linking agent, silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent, silane reinforcer, silylating agent, silane reducing agent, silyl building blocks and synthons.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Chemical Identifiers
17980-47-1 | |
220.384 | |
ALVYUZIFSCKIFP-UHFFFAOYSA-N | |
87391 | |
CCO[Si](CC(C)C)(OCC)OCC |
C10H24O3Si | |
MFCD00069084 | |
triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilaneShow More | |
triethoxy(2-methylpropyl)silane |
Specifications
165°C (400mmHg) | |
17980-47-1 | |
C10H24O3Si | |
MFCD00069084 | |
triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilane | |
ALVYUZIFSCKIFP-UHFFFAOYSA-N | |
triethoxy(2-methylpropyl)silane | |
87391 | |
95% | |
0.880 g/mL |
190°C to 191°C | |
Liquid | |
(CH3)2CHCH2Si(OC2H5)3 | |
1750049 | |
Insoluble in water. | |
CCO[Si](CC(C)C)(OCC)OCC | |
220.384 | |
220.38 | |
1.4505 | |
Isobutyltriethoxysilane |
Safety and Handling
GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'einecsNumber' : 402-810-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.