missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Acid, 99+%, Thermo Scientific Chemicals

Product Code. 10356470
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-31-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002658
InChI Key KQNPFQTWMSNSAP-UHFFFAOYSA-N
Synonym isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
PubChem CID 6590
ChEBI CHEBI:16135
IUPAC Name 2-methylpropanoic acid
SMILES CC(C)C(O)=O

Specifications

Melting Point -47.0°C
Boiling Point 153.0°C to 154.0°C
Color Colorless
Assay Percent Range 99+%
Linear Formula (CH3)2CHCO2H
Beilstein 02, 288
Merck Index 15, 5200
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: miscible with alcohol, chloroform and ether
Formula Weight 88.11
Percent Purity ≥99%
Density 0.9500g/mL
Quantity 1 kg
Flash Point 59°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3920 to 1.3940
Specific Gravity 0.95
Chemical Name or Material Isobutyric acid, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutyric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.