missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%, Thermo Scientific Chemicals

Product Code. 11303536
Click to view available options
Quantity:
250 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001873
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
PubChem CID 6559
ChEBI CHEBI:28638
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

Melting Point -72°C
Density 0.77
Boiling Point 103°C to 104°C
Flash Point 8°C (46°F)
Linear Formula (CH3)2CHCN
Refractive Index 1.374
Quantity 1000 mL
UN Number UN2284
Beilstein 1340512
Merck Index 14,5156
Solubility Information Soluble in water 35g/L (20°C).
Formula Weight 69.11
Percent Purity 99%
Chemical Name or Material Isobutyronitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'