Learn More
Isobutyryl chloride, 98%, Thermo Scientific Chemicals
£23.40 - £119.00
Chemical Identifiers
CAS | 79-30-1 |
---|---|
Molecular Formula | C4H7ClO |
Molecular Weight (g/mol) | 106.55 |
MDL Number | MFCD00000717 |
InChI Key | DGMOBVGABMBZSB-UHFFFAOYSA-N |
Synonym | isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride |
PubChem CID | 62325 |
IUPAC Name | 2-methylpropanoyl chloride |
SMILES | CC(C)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10583521
|
Thermo Scientific Acros
160660100 |
10 mL | Glass bottle |
£23.40
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10255730
|
Thermo Scientific Acros
160662500 |
250 mL | Glass bottle |
£45.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10275680
|
Thermo Scientific Acros
160660010 |
1 L | Glass bottle |
£119.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-30-1 | |
106.55 | |
DGMOBVGABMBZSB-UHFFFAOYSA-N | |
62325 | |
CC(C)C(=O)Cl |
C4H7ClO | |
MFCD00000717 | |
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride | |
2-methylpropanoyl chloride |
Specifications
79-30-1 | |
1.0170g/mL | |
1°C | |
97.5% min. (GC) | |
(CH3)2CHCOCl | |
MFCD00000717 | |
1.017 | |
Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether | |
CC(C)C(=O)Cl | |
106.55 | |
0.55 mPa.s (20°C) | |
98% | |
Isobutyryl chloride, 98% |
-90.0°C | |
91.0°C to 93.0°C | |
Authentic | |
C4H7ClO | |
1.4060 to 1.4080 | |
02, 293 | |
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride | |
DGMOBVGABMBZSB-UHFFFAOYSA-N | |
2-methylpropanoyl chloride | |
62325 | |
106.55 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Fatal if inhaled.
May be corrosive to metals.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-194-1
missing translation for 'rtecsNumber' : UC3944000
missing translation for 'tsca' : TSCA
RUO – Research Use Only