missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck Product Code.: 15644914
Add to basket

DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
BIOPHARMA

Product Code. 15644914 Shop All DuPont Products
Medium
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 15644914

Brand: DuPont™ D15466127

Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting.

  • Available and in sizes S to XXXL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunneled elastication at wrists.
  • Front snap closure for easy donning and doffing.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Lab Coat
Size Medium
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Tunnelized Elastic
Disposable Single-use
Sterility Sterile
Collar Type Bound
Seam Type Bound internal seams
Size (Metric) Chest 93 to 102 cm
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Full traceability on all sterilized apparel with certificates of sterility available, Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
For Use With (Application) Cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Show More Show Less
  • Dupont Tyvek - IsoClean design
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Dupont Tyvek IsoClean design

  • Dupont Tyvek - Packaging for CE certified IsoClean garments
    Video Player is loading.
    Current Time 0:00
    Duration 1:40
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:40
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Dupont Tyvek Packaging for CE certified IsoClean garments

  • DuPont Tyvek - IsoClean
    Video Player is loading.
    Current Time 0:00
    Duration 2:33
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:33
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    DuPont Tyvek IsoClean

Brochures

Posters

Product Information

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAHS2414138 Certificate TypeCertificate of Sterility Date20/07/2024 Catalog NumberD15466127
Lot NumberAHS2413112 Certificate TypeCertificate of Sterility Date20/07/2024 Catalog NumberD15466127
Lot NumberAHS2403177 Certificate TypeCertificate of Sterility Date05/04/2024 Catalog NumberD15466127
Lot NumberAHS2409141 Certificate TypeCertificate of Sterility Date05/04/2024 Catalog NumberD15466127
Lot NumberAHS2350166 Certificate TypeCertificate of Sterility Date18/01/2024 Catalog NumberD15466127

The user should make the risk assessment to determine the protection required.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.