missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck Product Code.: 15674914
Add to basket

DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
BIOPHARMA

Product Code. 15674914 Shop All DuPont Products
2X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 15674914

Brand: DuPont™ D15466157

Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting.

  • Available and in sizes S to XXXL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunneled elastication at wrists.
  • Front snap closure for easy donning and doffing.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Tunnelized Elastic
Disposable Single-use
Sterility Sterile
Collar Type Bound
Seam Type Bound internal seams
Size (Metric) Chest 114 to 123 cm
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Full traceability on all sterilized apparel with certificates of sterility available, Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
For Use With (Application) Cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Mehr anzeigen Weniger anzeigen
  • Dupont Tyvek - Packaging for CE certified IsoClean garments
    Video Player is loading.
    Current Time 0:00
    Duration 1:40
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:40
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Dupont Tyvek Packaging for CE certified IsoClean garments

  • Dupont Tyvek - IsoClean design
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Dupont Tyvek IsoClean design

    • DuPont Tyvek - IsoClean
      Video Player is loading.
      Current Time 0:00
      Duration 2:33
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 2:33
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected

      DuPont Tyvek IsoClean

    Brochures

    Posters

    Product Information

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberAHS2448139 Certificate TypeCertificate of Sterility Date13/02/2025 Catalog NumberD15466157
    Lot NumberAHS2444131 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466157
    Lot NumberAHS2437152 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466157
    Lot NumberAHS2235144 Certificate TypeCertificate of Sterility Date28/09/2022 Catalog NumberD15466157
    Lot NumberAHS1625419 Certificate TypeCertificate of Sterility Date21/09/2022 Catalog NumberD15466157

    The user should make the risk assessment to determine the protection required.

    Berichtigung von Produktinhalten

    Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.

    Name des Produkts
    DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck >

    Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

    Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde ĂĽbermittelt