Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting.
ÂŁ304.21 - ÂŁ313.16
Specifications
| Product Type | Controlled Environments Lab Coat |
|---|---|
| Material | Tyvek |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Tunnelized Elastic |
| Product Code | Brand | Size | Price | Quantity & Availability | ||||||
|---|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15634914
![]() |
DuPont™
D15466113 |
Small |
ÂŁ379.00 ÂŁ304.21 / Pack of 30 Save ÂŁ74.79 19% Off |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||||
15644914
![]() |
DuPont™
D15466127 |
Medium |
ÂŁ379.00 ÂŁ304.21 / Pack of 30 Save ÂŁ74.79 19% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
||||||
15654914
![]() |
DuPont™
D15466137 |
Large |
ÂŁ379.00 ÂŁ304.21 / Pack of 30 Save ÂŁ74.79 19% Off |
Estimated Shipment: 27-11-2025
Log in to see stock. |
||||||
15664914
![]() |
DuPont™
D15466146 |
X-Large |
ÂŁ394.00 ÂŁ304.21 / Pack of 30 Save ÂŁ89.79 22% Off |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||||
15674914
![]() |
DuPont™
D15466157 |
2X-Large |
ÂŁ384.00 ÂŁ304.21 / Pack of 30 Save ÂŁ79.79 20% Off |
Estimated Shipment: 27-11-2025
Log in to see stock. |
||||||
15684914
![]() |
DuPont™
D15466164 |
3X-Large |
ÂŁ390.00 ÂŁ313.16 / Pack of 30 Save ÂŁ76.84 19% Off |
Estimated Shipment: 04-12-2025
Log in to see stock. |
||||||
Description
- Available and in sizes S to XXXL.
- Clean-processed and gamma-sterilized.
- Bound internal seams.
- Tunneled elastication at wrists.
- Front snap closure for easy donning and doffing.
Specifications
| Controlled Environments Lab Coat | |
| White | |
| Tunnelized Elastic | |
| Sterile | |
| Bound internal seams | |
| Cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments. |
| Tyvek | |
| Snaps | |
| Single-use | |
| Bound | |
| Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Full traceability on all sterilized apparel with certificates of sterility available, Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I |
The user should make the risk assessment to determine the protection required.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.