missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Sterile Hood with Ties Product Code.: 15684904
Estimated Shipment: 09-04-2025
to see stock.
Add to basket

DuPont™ Tyvek™ IsoClean™ Sterile Hood with Ties
BIOPHARMA

Product Code. 15684904 Shop All DuPont Products
Size: Universal
Click to view available options
Size: Universal
This item is not returnable. View return policy

Product Code. 15684904

Brand: DuPont™ D15466063

Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and have been sterilized by gamma- irradiation. DuPont™ Tyvek™ IsoClean™ Sterile Hood with Ties provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and low linting.

  • Clean-processed and gamma-sterilized
  • Bound seams
  • Bound hood opening for low particle shedding
  • Full face opening
  • Ties with loops for adjustable fit
  • White, one size
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hood with ties
Size Universal
Material Flashspun HDPE
Color White
Closure Type Ties with Loops
Disposable Single-use
Model D15466063
Seam Type Bound
Sterility Sterile
Certifications/Compliance ISO 11137-1, PPE Category I
Packaging Type Individually Packaged
Quantity 100/Pk.
For Use With (Application) Cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Hood Type Bound
Show More Show Less
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Dupont Tyvek Packaging for CE certified IsoClean garments

    • Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Dupont Tyvek IsoClean design

      • Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
         
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected

          DuPont Tyvek IsoClean

        Brochures

        Posters

        Product Information

        Certificates

        A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

        5 results found
        Lot Number Certificate Type Date Catalog Number
        Lot NumberAHS2447146 Certificate TypeCertificate of Sterility Date17/12/2024 Catalog NumberD15466063
        Lot NumberAHS2445152 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466063
        Lot NumberAHS2441139 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466063
        Lot NumberAHS2441123 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466063
        Lot NumberAHS2444130 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466063

        The user should make the risk assessment to determine the protection required.

        Product Content Correction

        Your input is important to us. Please complete this form to provide feedback related to the content on this product.

        Product Title
        DuPont™ Tyvek™ IsoClean™ Sterile Hood with Ties >

        By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

        Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.