missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Boot Covers
BIOPHARMA

Product Code. 15604914
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. Dupont™ Tyvek™ IsoClean™ Boot Covers provide an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting

  • Available in sizes S to XL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunnelled elastication at shin and ankles.
  • Ankle ties.
  • Gripper™ soles provide enhanced slip-retardance and robustness.
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Tyvek™ IsoClean™
Color White
Disposable Single-use
Sterility Sterile
Sole Type Anti-skid
Seam Type Bound
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics, Other Critical Environments
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
Packaging Quantity 100/Cs.
Packaging Type Individually Packaged in Pairs
Product Type Controlled Environments Boot Covers
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.