missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers
Shop All DuPont Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
Clean-processed and gamma-sterilized. DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash.
- Hooded coverall includes attached slip-retardant shoe covers
- Clean-processed and gamma-sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
- Serged internal seams
- Elastication at wrists, ankles and face
- Thumb loops prevent sleeves from riding up
- Zipper closure
- Aseptically folded
- White.

Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | X-Large |
Material | HDPE |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Front Zipper |
Seam Type | Serged |
Show More |
Videos
-
Dupont Tyvek Packaging for CE certified IsoClean garments
-
Dupont Tyvek IsoClean design
-
DuPont Tyvek IsoClean
Documents
Scientific Resources
Brochures
IsoClean
File size: 1.5 MB
Posters
Clenroom gowning Poster
File size: 516.2 kB
Product Information
Product Information
IsoClean FAQ
File size: 122.3 kB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2447142 | Certificate of Sterility | 17/12/2024 | D14247990 |
AHS2445128 | Certificate of Sterility | 11/12/2024 | D14247990 |
AHS2446141 | Certificate of Sterility | 11/12/2024 | D14247990 |
AHS2446123 | Certificate of Sterility | 11/12/2024 | D14247990 |
AHS2445156 | Certificate of Sterility | 11/12/2024 | D14247990 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers >
Spot an opportunity for improvement?Share a Content Correction