missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isododecane, mixture of isomers, tech. 80%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Density | 0.74 |
Boiling Point | 170°C to 195°C |
Flash Point | 42°C (108°F) |
Assay Percent Range | 80% |
Refractive Index | 1.421 |
Quantity | 500 mL |
UN Number | UN3295 |
Solubility Information | Immiscible with water. |
Formula Weight | 170.34 |
Percent Purity | 80% |
Show More |
Safety and Handling
Product Identifier
- Isododecane
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P331-Do NOT induce vomiting.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : Isododecane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'