Learn More
Isoeugenol, 98+%, mixture of cis/trans isomers, Thermo Scientific Chemicals
£13.40 - £109.00
Chemical Identifiers
CAS | 97-54-1 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.20 |
MDL Number | MFCD00009285 |
InChI Key | BJIOGJUNALELMI-ONEGZZNKSA-N |
Synonym | isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene |
PubChem CID | 853433 |
ChEBI | CHEBI:50545 |
SMILES | COC1=CC(\C=C\C)=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10284970
|
Thermo Scientific Acros
122570050 |
5 g | Glass bottle |
£13.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783681
|
Thermo Scientific Acros
122571000 |
100 g | Glass bottle |
£30.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10152920
|
Thermo Scientific Acros
122575000 |
500 g | Glass bottle |
£109.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-54-1 | |
164.20 | |
BJIOGJUNALELMI-ONEGZZNKSA-N | |
853433 | |
COC1=CC(\C=C\C)=CC=C1O |
C10H12O2 | |
MFCD00009285 | |
isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene | |
CHEBI:50545 |
Specifications
97-54-1 | |
Yellow | |
266.0°C to 268.0°C | |
Authentic | |
C10H12O2 | |
1.5760 to 1.5780 | |
06,II,916 | |
1.09 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether and other organic,solvents | |
COC1=CC(\C=C\C)=CC=C1O | |
164.20 | |
CHEBI:50545 | |
164.2 | |
Viscous Liquid |
19.0°C to 23.0°C | |
1.0900g/mL | |
150°C | |
98% min. (GC) | |
CH3OC6H3(CH=CHCH3)OH | |
MFCD00009285 | |
15, 5216 | |
isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene | |
BJIOGJUNALELMI-ONEGZZNKSA-N | |
2-methoxy-4-[(1E)-prop-1-en-1-yl]phenol | |
853433 | |
26.7 mPa.s (25°C) | |
98+% | |
Isoeugenol, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-590-7
missing translation for 'rtecsNumber' : SL7875000
missing translation for 'tsca' : TSCA
- Isoeugenol, mixture of cis/trans isomers
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only