missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isohexane, contains <5% N-Hexane, Certified AR for Analysis
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Metal Can
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | -154°C |
Color | Colorless |
Boiling Point | 60°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Formula Weight | 86.18g/mol |
Safety and Handling
Product Identifier
- Isohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Hydrocarbons, C6, isoalkanes < 5% n-hexane (Iso-Hexane), Hexane, mixed isomers
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'