Learn More
Thermo Scientific™ Isolator™ 10 Tube Accessories
Support the functionality of Thermo Scientific™ Oxoid™ Isolator™ 10 Tubes Blood Culture System with using these accessories.
Brand: Thermo Scientific™ BC0507C
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Support the functionality of Thermo Scientific™ Isolator™ 10 Tubes Blood Culture System with Isostat™ Accessories. The Isolator 10 Tubes Blood Culture System is a highly sensitive blood culture system for the rapid detection and isolation of microorganisms in a single process.
Support the functionality of Isolator 10 Tubes Blood Culture System with Isostat Accessories.
Isolator 10 Tube Blood Culture System provides simple, rapid and sensitive method for detection of bacteraemia.1
- Rapid detection: the Isolator tubes combine detection and isolation in a single process making isolated colonies available 30 hours earlier than with traditional methods1.
- Superior sensitivity: the Isolator blood culture system has recovery rates comparable to, or better than, conventional broth techniques1,2,3
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Isostat™ Press and Rack | |
Isostat™ Press and Rack | |
Each |