Learn More
Thermo Scientific™ Isolator Wrap Sterile Settle Tryptic Soy Agar Plates with Lecithin & Polysorbate 80
Thermo Scientific™ Remel Sterile Settling Plate Tryptic Soy Agar is used for the cultivation of bacteria, yeast and fungi.
Brand: Thermo Scientific™ R111815
88.46 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.60000kg
Description
- VHP resistant packaged Settling plates (single bag and double bag) are manufactured in a clean room environment. This provides the highest level of sterility assurance with no compromise of growth characteristics with 23 g fill volume.
- Each lot is tested to ensure consistent, reproducible results.
- Storage Condition: 2 to 8°C.
Specifications
2°C to 8°C, Refrigerated | |
Isolator Settling Plate | |
10/Pk. |
Tryptic Soy Agar | |
RABS, Isolators and Cleanrooms | |
85 mm |