missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
It acts as an useful isomer of nicotinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as an useful isomer of nicotinic acid.
Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.
Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It acts as an useful isomer of nicotinic acid.
Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.
Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | ∼310°C (sublimation) |
pH | 3.0 to 4.0 |
Boiling Point | 260°C (15 mmHg) |
Quantity | 500 g |
Beilstein | 109599 |
Merck Index | 14,5187 |
Solubility Information | Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol. |
Formula Weight | 123.11 |
Percent Purity | 99% |
Chemical Name or Material | Isonicotinic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252577 | Certificate of Analysis | 26/09/2024 | A18109.0E,A18109.36,A18109.22 |
10245123 | Certificate of Analysis | 24/10/2023 | A18109.0E,A18109.36,A18109.22 |
10240886 | Certificate of Analysis | 26/05/2022 | A18109.0E,A18109.36,A18109.22 |
5007P25T | Certificate of Analysis | 23/11/2021 | A18109.0E,A18109.36,A18109.22 |
GBFA005791 | Certificate of Analysis | 23/11/2021 | A18109.0E,A18109.36,A18109.22 |
Safety and Handling

Product Identifier
- Isonicotinic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Isonicotinic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Isonicotinic acid, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction