missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic anhydride, 90%, remainder picolinic and nicotinic acid-anhydride, Thermo Scientific Chemicals

Product Code. 10287060
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7082-71-5
Melting Point 133°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C12H8N2O3
MDL Number MFCD00062784
Quantity 5 g
Synonym isonicotinic anhydride, isonictinic anhydride, acmc-1bcxw, isonicotinic acid anhydride, bis 4-pyridinecarboxylic anhydride, 4-pyridinecarboxylicacid, anhydride with 4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid anhydride; pyridine-4-carboxylic acid anhydride
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: hydrolysis in water
InChI Key ORBIGZZHMAJXAZ-UHFFFAOYSA-N
SMILES C1=CN=CC=C1C(=O)OC(=O)C2=CC=NC=C2
IUPAC Name pyridine-4-carbonyl pyridine-4-carboxylate
Molecular Weight (g/mol) 228.2
PubChem CID 81529
Formula Weight 228.2
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material Isonicotinic anhydride, 90%
Show More Show Less
compliance-icons
Product Identifier
  • Isonicotinic anhydride, remainder picolinic and nicotinic acid-anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'