missing translation for 'onlineSavingsMsg'
Learn More

Isopentyltriphenylphosphonium bromide, 98%, Thermo Scientific™

Product Code. 11433724
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28322-40-9
Molecular Formula C23H26BrP
Molecular Weight (g/mol) 413.34
MDL Number MFCD00031594
InChI Key GZLGTVRDLCJQTO-UHFFFAOYSA-M
Synonym isopentyltriphenylphosphonium bromide, isoamyltriphenylphosphonium bromide, isoamyl triphenylphosphonium bromide, 3-methylbutyl triphenylphosphanium bromide, 3-methylbutyl triphenylphosphonium bromide, 3-methylbutyl triphenyl phosphanium bromide, 2-isopropoxyethyl triphenylphosphonium bromide, acmc-1cjq9, isoamyltriphenylphosphoniumbromide, iso-amyl triphenylphosphonium bromide
PubChem CID 2759379
SMILES [Br-].CC(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼155°C
Quantity 250 g
Beilstein 3804221
Sensitivity Hygroscopic
IUPAC Name 3-methylbutyl(triphenyl)phosphanium;bromide
Formula Weight 413.35
Percent Purity 98%
Chemical Name or Material Isopentyltriphenylphosphonium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'