Learn More
Isophorone, 97%, Thermo Scientific Chemicals
£11.98 - £42.70
Chemical Identifiers
CAS | 78-59-1 |
---|---|
Molecular Formula | C9H14O |
Molecular Weight (g/mol) | 138.21 |
MDL Number | MFCD00001584 |
InChI Key | HJOVHMDZYOCNQW-UHFFFAOYSA-N |
Synonym | isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one |
PubChem CID | 6544 |
ChEBI | CHEBI:34800 |
IUPAC Name | 3,5,5-trimethylcyclohex-2-en-1-one |
SMILES | CC1=CC(=O)CC(C)(C)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10518860
|
Thermo Scientific Acros
122640050 |
5 mL | Glass bottle | ||||||
10021210
|
Thermo Scientific Acros
122645000 |
500 mL | Glass bottle |
£20.40
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10550971
|
Thermo Scientific Acros
122640010 |
1 L | Glass bottle |
£31.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13478328
|
Thermo Scientific Acros
122640025 |
2.5 L | Glass bottle |
£42.70
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-59-1 | |
138.21 | |
HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
6544 | |
3,5,5-trimethylcyclohex-2-en-1-one |
C9H14O | |
MFCD00001584 | |
isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
CHEBI:34800 | |
CC1=CC(=O)CC(C)(C)C1 |
Specifications
78-59-1 | |
Colorless to Yellow | |
213.0°C to 214.0°C | |
Authentic | |
C9H14O | |
MFCD00001584 | |
15,5242 | |
isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one | |
HJOVHMDZYOCNQW-UHFFFAOYSA-N | |
3,5,5-trimethylcyclohex-2-en-1-one | |
6544 | |
2.6 mPa.s (20°C) | |
97% | |
Isophorone, 98% |
-8.0°C | |
0.9200g/mL | |
84°C | |
96% min. (GC) | |
1.4770 to 1.4790 | |
07,65 | |
0.92 | |
Solubility in water: 12g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone | |
CC1=CC(=O)CC(C)(C)C1 | |
138.21 | |
CHEBI:34800 | |
138.21 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Suspected of causing cancer.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-126-0
missing translation for 'rtecsNumber' : GW7700000
missing translation for 'tsca' : TSCA
- Isophorone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only