Learn More
Isophorone diisocyanate, 98%, Thermo Scientific Chemicals
£19.10 - £158.00
Chemical Identifiers
CAS | 4098-71-9 |
---|---|
Molecular Formula | C12H18N2O2 |
Molecular Weight (g/mol) | 222.29 |
MDL Number | MFCD00064956 |
InChI Key | NIMLQBUJDJZYEJ-UHFFFAOYNA-N |
Synonym | isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates |
PubChem CID | 169132 |
ChEBI | CHEBI:53214 |
IUPAC Name | 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane |
SMILES | CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10555290
|
Thermo Scientific Acros
427600500 |
50 mL | Glass bottle |
£19.10
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10844161
|
Thermo Scientific Acros
427602500 |
250 mL | Glass bottle |
£56.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646640
|
Thermo Scientific Acros
427600010 |
1 L | Glass bottle |
£158.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4098-71-9 | |
222.29 | |
NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
169132 | |
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane |
C12H18N2O2 | |
MFCD00064956 | |
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
CHEBI:53214 | |
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O |
Specifications
4098-71-9 | |
1.0610g/mL | |
163°C | |
97.5% min. (sum of mixed isomers) (GC) | |
1.4830 to 1.4850 (20°C, 589nm) | |
1.061 | |
NIMLQBUJDJZYEJ-UHFFFAOYNA-N | |
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane | |
169132 | |
13-15 mPa.s (23°C) | |
98% | |
Isophorone diisocyanate |
Colorless to Yellow | |
153.0°C (10.0 mmHg) | |
Authentic | |
C12H18N2O2 | |
MFCD00064956 | |
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates | |
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O | |
222.29 | |
CHEBI:53214 | |
222.28 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic to aquatic life with long lasti
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-861-6
- Isophorone diisocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H331-Toxic if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only