Learn More
Merck Isopore™ Membrane Filters, 0.6 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
£128.00 - £239.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Bubble Point | 8.5 psi |
Color | White |
Filter Type | Screen Filter |
Material | Polycarbonate |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10298150
|
Merck
DTTP01300 |
13 mm |
£128.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
10733291
|
Merck
DTTP02500 |
25 mm |
£203.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
10278010
|
Merck
DTTP04700 |
47 mm |
£239.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
White | |
Polycarbonate | |
Non-sterile | |
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. | |
Hydrophilic | |
100/Pk. | |
Hydrophilic |
8.5 psi | |
Screen Filter | |
0.6 μm | |
Avg. 43 mL/min x cm2 (typical results (at 10 psi);62 mL/min x cm2/psi) | |
10.0 L/min/cm2 | |
100Pack | |
Filter Membranes |