Learn More
Merck Isopore™ Membrane Filters, 10 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
£238.00 - £712.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen Filter |
Material | Polycarbonate |
Pore Size | 10 μm |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10627321
|
Merck
TCTP01300 |
13 mm |
£238.00
Pack of 100 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
10517492
|
Merck
TCTP02500 |
25 mm |
£302.00
Pack of 100 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
10627511
|
Merck
TCTP04700 |
47 mm |
£456.00
Pack of 100 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10005391
|
Merck
TCTP14250 |
142 mm |
£712.00
Pack of 50 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
Screen Filter | |
10 μm | |
Avg. 900 mL/min x cm2 (typical results (at 10 psi)) | |
69.0 L/min/cm2 | |
Filter Membranes |
White | |
Polycarbonate | |
Non-sterile | |
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. | |
Hydrophilic | |
Hydrophilic |