Learn More
Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals
£93.87 - £271.00
Chemical Identifiers
CAS | 51-30-9 |
---|---|
Molecular Formula | C11H18ClNO3 |
Molecular Weight (g/mol) | 247.72 |
MDL Number | MFCD00012603,MFCD00064548 |
InChI Key | IROWCYIEJAOFOW-UHFFFAOYNA-N |
Synonym | isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso |
PubChem CID | 5807 |
IUPAC Name | 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride |
SMILES | [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11915431
|
Thermo Scientific Acros
437210250 |
25 g |
£93.87
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11925431
|
Thermo Scientific Acros
437211000 |
100 g |
£271.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-30-9 | |
247.72 | |
IROWCYIEJAOFOW-UHFFFAOYNA-N | |
5807 | |
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 |
C11H18ClNO3 | |
MFCD00012603,MFCD00064548 | |
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride |
Specifications
51-30-9 | |
1% max. | |
97.5% min. (HPLC) | |
C11H18ClNO3 | |
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
IROWCYIEJAOFOW-UHFFFAOYNA-N | |
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride | |
5807 | |
98% |
165.0°C to 175.0°C | |
Authentic | |
Glass bottle | |
MFCD00012603,MFCD00064548 | |
Solubility in water: soluble in cold water. | |
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 | |
247.72 | |
247.72 | |
Isoprenaline hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-089-8
- Isoprenaline hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Isoprenaline hydrochloride
RUO – Research Use Only