missing translation for 'onlineSavingsMsg'
Learn More
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals Product Code.: 10723124

Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Product Code. 10723124
250 mL, Glass Bottle
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10723124

Brand: Thermo Scientific Acros 327272500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petroholShow MoreShow Less
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 2.1 (Air = 1.0)
Quantity 250 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5%
Residue after Evaporation 0.0005% max.
Linear Formula (CH3)2CHOH
Vapor Pressure Vapor Pressure: 43mmHg at 20°C
Beilstein 01, 360
Water 0.05% max. (Coulometric)
Fieser 13,108
Merck Index 15, 5254
Formula Weight 60.1
Percent Purity ≥99.5%
Grade Molecular Biology
Chemical Name or Material Isopropanol, DNAse, RNAse and Protease Free
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463496 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number327272500,327270010
Lot NumberA0464840 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number327272500,327270010
Lot NumberA0463496 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number327272500,327270010
Lot NumberA0462179 Certificate TypeCertificate of Analysis Date16/07/2024 Catalog Number327272500,327270010
Lot NumberA0459779 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number327272500,327270010
compliance-icons compliance-icons
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.