Learn More
Isopropanol, Certified AR for Analysis, Fisher Chemical™
Description
C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096

Specifications
Specifications
pH | 7 |
Melting Point | -89.5°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 2.5 L |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Vapor Pressure | 43mmHg at 20°C |
Viscosity | 2.27 mPaS at 20°C |
Formula Weight | 60.1g/mol |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling


- Propan-2-ol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Isopropyl alcohol
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
34Hours
17Minutes
9Seconds
31New Offer Save Up to 70% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.