missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Optima™LC/MS Grade, Fisher Chemical™

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

£43.05 - £169.00

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10783447
View Documents
Fisher Chemical
A461-500
500 mL Glass Bottle
£43.05
500mL
Please to purchase this item. Need a web account? Register with us today!
10091304
View accessory products View Documents
Fisher Chemical
A461-1
1 L Wide Neck Amber Bottle
£66.10
1 L
Please to purchase this item. Need a web account? Register with us today!
10684355
View accessory products View Documents Promotion Details
Fisher Chemical
A461-212
2.5 L Glass Bottle
£130.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10001314
View Documents Promotion Details
Fisher Chemical
A461-4
4 L Glass Bottle
£169.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
11674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

7
Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Colorless
81°C
Assay: ≥99.90%
C3H8O
11674
2.27 mPaS at 20°C
15, 5254
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
LC-MS & UHPLC-UV
-89.5°C
67-63-0
0.785g/cm³
Liquid
1ppm max.
(CH3)2CHOH
43mmHg at 20°C
0.05% max.
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.90%
2-Propanol
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

Product Identifier
  • Iso-propanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.