missing translation for 'onlineSavingsMsg'
Learn More

Isopropenyl acetate, 99%, Thermo Scientific Chemicals

Product Code. 10337950
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-22-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008709
InChI Key HETCEOQFVDFGSY-UHFFFAOYSA-N
Synonym isopropenyl acetate, 2-acetoxypropene, 1-methylvinyl acetate, 2-acetoxypropylene, methylvinyl acetate, 1-propen-2-ol, acetate, propen-2-yl acetate, acetic acid isopropenyl ester, 1-acetoxy-1-methylethylene, 1-propen-2-yl acetate
PubChem CID 7916
IUPAC Name prop-1-en-2-yl acetate
SMILES CC(=C)OC(C)=O

Specifications

Melting Point -93°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 94°C to 97°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2C(CH3)=CH2
Refractive Index 1.4000 to 1.402
Quantity 1 kg
Beilstein 02, II, 278
Fieser 01,524; 04,273; 06,308
Merck Index 15, 5249
Specific Gravity 0.92
Solubility Information Solubility in water: 34g/L in water (20°C). Other solubilities: miscible with nearly all organic solvents
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropenyl acetate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'