Learn More
Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
£76.60 - £322.00
Chemical Identifiers
CAS | 13291-18-4, 109-99-9 |
---|---|
Molecular Formula | C3H5BrMg |
Molecular Weight (g/mol) | 145.28 |
MDL Number | MFCD00192240 |
InChI Key | LFTYALBVGVNGLI-UHFFFAOYSA-M |
Synonym | isopropenylmagnesium bromide, isopropenylmagnesium bromide solution, isopropenyl magnesium bromide, bromo prop-1-en-2-yl magnesium, magnesium, bromo 1-methylethenyl, bromo isopropenyl magnesium, 1-methylethenylmagnesium bromide, 2-propen-2-ylmagnesium bromide, prop-1-en-2-ylmagnesium bromide, lftyalbvgvngli-uhfffaoysa-m, grignard reagent |
PubChem CID | 4187787 |
IUPAC Name | magnesium;prop-1-ene;bromide |
SMILES | CC(=C)[Mg]Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10112774
|
Thermo Scientific Acros
427751000 |
100 mL | AcroSeal™ Glass bottle |
£76.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10684175
|
Thermo Scientific Acros
427758000 |
800 mL | AcroSeal™ Glass Bottle |
£322.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13291-18-4, 109-99-9 | |
145.28 | |
LFTYALBVGVNGLI-UHFFFAOYSA-M | |
4187787 | |
CC(=C)[Mg]Br |
C3H5BrMg | |
MFCD00192240 | |
isopropenylmagnesium bromide, isopropenylmagnesium bromide solution, isopropenyl magnesium bromide, bromo prop-1-en-2-yl magnesium, magnesium, bromo 1-methylethenyl, bromo isopropenyl magnesium, 1-methylethenylmagnesium bromide, 2-propen-2-ylmagnesium bromide, prop-1-en-2-ylmagnesium bromide, lftyalbvgvngli-uhfffaoysa-m, grignard reagent | |
magnesium;prop-1-ene;bromide |
Specifications
13291-18-4, 109-99-9 | |
0.9270g/mL | |
2°C | |
CH2=C(CH3)MgBr | |
0.927 | |
LFTYALBVGVNGLI-UHFFFAOYSA-M | |
magnesium;prop-1-ene;bromide | |
4187787 | |
Crystalline Powder |
Orange to White | |
67.0°C | |
C3H5BrMg | |
MFCD00192240 | |
isopropenylmagnesium bromide, isopropenylmagnesium bromide solution, isopropenyl magnesium bromide, bromo prop-1-en-2-yl magnesium, magnesium, bromo 1-methylethenyl, bromo isopropenyl magnesium, 1-methylethenylmagnesium bromide, 2-propen-2-ylmagnesium bromide, prop-1-en-2-ylmagnesium bromide, lftyalbvgvngli-uhfffaoysa-m, grignard reagent | |
CC(=C)[Mg]Br | |
145.28 | |
145.28 | |
Isopropenylmagnesium bromide, 0.5M Solution in THF |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May form explosive
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
- Isopropenylmagnesium bromide, 05M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only