missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Description
It is used as a preservative in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a preservative in cosmetics.
Solubility
Soluble in Chloroform, Ethyl Acetate.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
It is used as a preservative in cosmetics.
Solubility
Soluble in Chloroform, Ethyl Acetate.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Specifications
Specifications
Melting Point | 84°C to 86°C |
Quantity | 50 g |
Beilstein | 2641564 |
Solubility Information | Soluble in Chloroform,Ethyl Acetate. |
Formula Weight | 180.21 |
Percent Purity | 98% |
Chemical Name or Material | Isopropyl 4-hydroxybenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'