Learn More
Isopropyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
£18.10 - £371.00
Chemical Identifiers
CAS | 4191-73-5 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.20 |
MDL Number | MFCD00016468 |
InChI Key | CMHMMKSPYOOVGI-UHFFFAOYSA-N |
Synonym | isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester |
PubChem CID | 20161 |
IUPAC Name | propan-2-yl 4-hydroxybenzoate |
SMILES | CC(C)OC(=O)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498860
|
Thermo Scientific Alfa Aesar
A13930.06 |
5 g |
£18.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431658
|
Thermo Scientific Alfa Aesar
A13930.09 |
10 g |
£26.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441658
|
Thermo Scientific Alfa Aesar
A13930.18 |
50 g |
£94.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451658
|
Thermo Scientific Alfa Aesar
A13930.30 |
250 g |
£371.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a preservative in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a preservative in cosmetics.
Solubility
Soluble in Chloroform, Ethyl Acetate.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Chemical Identifiers
4191-73-5 | |
180.20 | |
CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
20161 | |
CC(C)OC(=O)C1=CC=C(O)C=C1 |
C10H12O3 | |
MFCD00016468 | |
isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
propan-2-yl 4-hydroxybenzoate |
Specifications
4191-73-5 | |
C10H12O3 | |
2641564 | |
Soluble in Chloroform,Ethyl Acetate. | |
CC(C)OC(=O)C1=CC=C(O)C=C1 | |
180.20 | |
180.21 | |
Isopropyl 4-hydroxybenzoate |
84°C to 86°C | |
MFCD00016468 | |
isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester | |
CMHMMKSPYOOVGI-UHFFFAOYSA-N | |
propan-2-yl 4-hydroxybenzoate | |
20161 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
missing translation for 'einecsNumber' : 224-069-3
missing translation for 'rtecsNumber' : DH2250000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only