Learn More
Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
£34.38 - £323.61
Chemical Identifiers
CAS | 108-21-4 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
MDL Number | MFCD00008877 |
InChI Key | JMMWKPVZQRWMSS-UHFFFAOYSA-N |
Synonym | isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl esterShow More |
PubChem CID | 7915 |
IUPAC Name | propan-2-yl acetate |
SMILES | CC(C)OC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10457550
|
Thermo Scientific Acros
150860010 |
1 L | Glass bottle |
£34.38
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10604632
|
Thermo Scientific Acros
150860025 |
2.5 L | Glass bottle |
£60.83
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10038571
|
Thermo Scientific Acros
150860250 |
25 L | Metal Drum |
£323.61
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-21-4 | |
102.13 | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
7915 | |
CC(C)OC(=O)C |
C5H10O2 | |
MFCD00008877 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl esterShow More | |
propan-2-yl acetate |
Specifications
108-21-4 | |
Colorless | |
88.8°C | |
Authentic | |
C5H10O2 | |
1.3760 to 1.3780 | |
02, 130 | |
0.872 | |
Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents | |
CC(C)OC(=O)C | |
102.13 | |
0.49 mPa.s (25°C) | |
99% | |
Liquid |
-73.0°C | |
0.8720g/mL | |
4°C | |
98.5% min. (GC) | |
CH3CO2CH(CH3)2 | |
MFCD00008877 | |
15, 5251 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
propan-2-yl acetate | |
7915 | |
102.13 | |
Pure | |
Isopropyl acetate, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-561-1
- Isopropyl acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.