Learn More
Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals
£20.20 - £205.02
Chemical Identifiers
CAS | 108-21-4 |
---|---|
MDL Number | MFCD00008877 |
InChI Key | JMMWKPVZQRWMSS-UHFFFAOYSA-N |
Synonym | isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester |
PubChem CID | 7915 |
IUPAC Name | propan-2-yl acetate |
SMILES | CC(C)OC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10439110
|
Thermo Scientific Acros
180801000 |
100 mL |
£20.20
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10082400
|
Thermo Scientific Acros
180800010 |
1 L |
£34.97
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10092400
|
Thermo Scientific Acros
|
2.5 L |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10667744
|
Thermo Scientific
180800250 |
25 L |
£205.02
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-21-4 | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
7915 | |
CC(C)OC(=O)C |
MFCD00008877 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester | |
propan-2-yl acetate |
Specifications
108-21-4 | |
Colorless | |
88.8°C | |
Authentic | |
C5H10O2 | |
1.3760 to 1.3780 | |
02, 130 | |
15, 5251 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
propan-2-yl acetate | |
7915 | |
≥99% | |
Liquid |
-73.0°C | |
0.8720g/mL | |
4°C | |
99+% | |
CH3CO2CH(CH3)2 | |
MFCD00008877 | |
0.01% max. | |
0.872 | |
Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents | |
CC(C)OC(=O)C | |
102.133 | |
102.13 | |
Pure | |
Isopropyl acetate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Isopropyl acetate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl acetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.