missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate, Puriss p.a., ≥99.5% (GC), Honeywell Riedel-de Haën™

Puriss. p.a.,≥99.5% (GC)

£44.51 - £104.03

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15685600
View Documents Promotion Details
Honeywell Riedel-de Haën
45960-250ML
250 mL Glass bottle
£44.51
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15675600
View Documents Promotion Details
Honeywell Riedel-de Haën
45960-1L
1 L Glass bottle
£104.03
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

108-21-4
102.133
JMMWKPVZQRWMSS-UHFFFAOYSA-N
7915
CC(C)OC(=O)C
C5H10O2
MFCD00008877
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
propan-2-yl acetate

Specifications

108-21-4
85°C
4°C (39.2°F)
C5H10O2
1.377
UN1220
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
CC(C)OC(=O)C
102.133
3.5 (vs air)
47mmHg (20°C)
Isopropyl acetate
0.872g/mL at 25°C
1.8% (37°F), 8%
894°F
CH3COOCH(CH3)2
MFCD00008877
1740761
JMMWKPVZQRWMSS-UHFFFAOYSA-N
propan-2-yl acetate
7915
102.13g/mol
≥99.5% (GC)
Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

missing translation for 'dotInformation' : class 3 - PG 2 - Isopropyl acetate

missing translation for 'rtecsNumber' : 203-561-1

SDS
Documents

Documents

Product Certifications