missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals
86.37 GBP valid until 2024-12-31
Use promo code "21958" to get your promotional price.
Use promo code "21958" to get your promotional price.
Click to view available options
Quantity:
500 mL
Specifications
Quantity | 500 mL |
Formula Weight | 60.09 |
Chemical Name or Material | Isopropyl Alcohol, 99% |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : isopropyl alcohol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'