missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
pH | Neutral |
Melting Point | -90°C |
Color | Colorless |
Linear Formula | (CH3)2CHOH |
UN Number | UN1219 |
Beilstein | 635639 |
Formula Weight | 60.1g/mol |
Density | 0.785g/mL at 25°C |
Boiling Point | 77°C to 83°C (1013 hPa) |
Quantity | 100 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'