missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl ether, 98+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Acidity | 0.005% max. |
Melting Point | -85.5°C |
Density | 0.7200g/mL |
Boiling Point | 68°C |
Flash Point | -29°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% min. (GC) |
Packaging | Metal Drum |
Linear Formula | [(CH3)2CH]2O |
Refractive Index | 1.3670 to 1.369 |
Show More |
Safety and Handling
Product Identifier
- Isopropyl ether
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- MIXTURE LIST-Contains : Diisopropyl ether
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'