Learn More
Isopropyl isocyanate, 98+%, Thermo Scientific Chemicals
Description
Isopropyl isocyanate is used as a reagent in the preparation of androgen receptor antagonist for prostate cancer treatment. It is also used in the synthesis of selective dual inhibitor of mammalian target of rapamycin (mTORC1 and mTORC2). Further, it serves as a reagent in the synthesis of O-aryl N-isopropylcarbamates. In addition to this, it is used as a derivatization reagent during the stereoisomeric analysis of secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Isopropyl isocyanate is used as a reagent in the preparation of androgen receptor antagonist for prostate cancer treatment. It is also used in the synthesis of selective dual inhibitor of mammalian target of rapamycin (mTORC1 and mT
Specifications
Specifications
CAS | 1795-48-8 |
Density | 0.868 |
Boiling Point | 74°C to 75°C |
Flash Point | −3°C (26°F) |
Odor | Acrid |
Molecular Formula | C4H7NO |
Refractive Index | 1.384 |
MDL Number | MFCD00002037 |
Quantity | 1 g |
UN Number | UN2483 |
Show More |
Safety and Handling
- Isopropyl isocyanate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Isocyanate disopropyle
RUO – Research Use Only