missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl isothiocyanate, 97%

Product Code. 11494743
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11494743

Brand: Thermo Scientific Alfa Aesar A11641.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2253-73-8 | C4H7NS | 101.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.

Solubility
Miscible with methanol. Hydrolyzes in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2253-73-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00037830
InChI Key VHBFEIBMZHEWSX-UHFFFAOYSA-N
Synonym isopropyl isothiocyanate, isothiocyanic acid isopropyl ester, isopropylisothiocyanate, propane, 2-isothiocyanato, unii-46561pkp9t, isopropyl mustard oil, 2-isothiocyanato-propane, propan-2-isothiocyanate, ipncs, isopropyl isothiocya
PubChem CID 75263
IUPAC Name 2-isothiocyanatopropane
SMILES CC(C)N=C=S

Specifications

Density 0.948
Boiling Point 137°C to 138°C
Flash Point 40°C (104°F)
Odor Pungent
Refractive Index 1.493
Quantity 5 g
UN Number UN2924
Beilstein 1739163
Sensitivity Moisture sensitive
Solubility Information Miscible with methanol. Hydrolyzes in water.
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Isopropyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.