missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl nitrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Isopropyl nitrate is used as a diesel cetane improver.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropyl nitrate is used as a diesel cetane improver.
Solubility
Partly miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, flammable substances, organic materials and metal powders.
Isopropyl nitrate is used as a diesel cetane improver.
Solubility
Partly miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, flammable substances, organic materials and metal powders.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.04 |
Boiling Point | 101°C to 102°C |
Flash Point | 12°C (54°F) |
Linear Formula | (CH3)2CHONO2 |
Refractive Index | 1.391 |
Quantity | 5 g |
UN Number | UN1222 |
Beilstein | 1701405 |
Solubility Information | Partly miscible in water. |
Formula Weight | 105.09 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'