missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine, 99+%, Thermo Scientific Chemicals

£17.49 - £73.26

Chemical Identifiers

CAS 75-31-0
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008082
InChI Key JJWLVOIRVHMVIS-UHFFFAOYSA-N
Synonym isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
PubChem CID 6363
ChEBI CHEBI:15739
IUPAC Name propan-2-amine
SMILES CC(C)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11417307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15044.AE
100 mL
£17.49
100mL
Please to purchase this item. Need a web account? Register with us today!
11427307
View Documents
Thermo Scientific Alfa Aesar
A15044.AP
500 mL
£23.62
500mL
Please to purchase this item. Need a web account? Register with us today!
11437307
View Documents
Thermo Scientific Alfa Aesar
A15044.0F
2500 mL
£73.26
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

Solubility
Miscible with water, chloroform, hexanes, acetone and ethanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and perchloryl fluoride.
Specifications

Chemical Identifiers

75-31-0
59.112
JJWLVOIRVHMVIS-UHFFFAOYSA-N
6363
propan-2-amine
C3H9N
MFCD00008082
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
CHEBI:15739
CC(C)N

Specifications

75-31-0
0.691
−32°C (−25°F)
Amine-like
(CH3)2CHNH2
MFCD00008082
605259
14,5209
Miscible with water,chloroform,hexanes,acetone and ethanol.
CC(C)N
59.112
CHEBI:15739
≥99%
-101°C
33°C to 34°C
≥99%
C3H9N
1.375
UN1221
Air Sensitive
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
JJWLVOIRVHMVIS-UHFFFAOYSA-N
propan-2-amine
6363
59.11
Isopropylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c

H224-H290-H301+H311+H331-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ISOPROPYLAMINE

missing translation for 'einecsNumber' : 200-860-9

missing translation for 'rtecsNumber' : NT8400000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Air sensitive

Recommended Storage : Ambient temperatures

Product Identifier
  • Isopropylamine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.