missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals
£39.60 - £108.54
Chemical Identifiers
CAS | 5306-85-4 |
---|---|
Molecular Formula | C8H14O4 |
Molecular Weight (g/mol) | 174.20 |
MDL Number | MFCD00064828 |
InChI Key | MEJYDZQQVZJMPP-UHFFFAOYNA-N |
Synonym | dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan |
PubChem CID | 62990 |
SMILES | COC1COC2C(COC12)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10764753
|
Thermo Scientific Acros
369910250 |
25 g | Glass bottle |
£39.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10272762
|
Thermo Scientific Acros
369911000 |
100 g | Glass bottle |
£108.54
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5306-85-4 | |
174.20 | |
MEJYDZQQVZJMPP-UHFFFAOYNA-N | |
62990 |
C8H14O4 | |
MFCD00064828 | |
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan | |
COC1COC2C(COC12)OC |
Specifications
5306-85-4 | |
1.1700g/mL | |
Authentic | |
1.4600 to 1.4620 | |
1.17 | |
MFCD00064828 | |
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan | |
MEJYDZQQVZJMPP-UHFFFAOYNA-N | |
3,6-dimethoxy-hexahydrofuro[3,2-b]furan | |
62990 | |
99% |
Colorless to Yellow | |
93°C to 95°C (0.1 hPa) | |
Liquid | |
98.5% min. (GC) | |
C8H14O4 | |
19, IV, 991 | |
Solubility in water: soluble. Other solubilities: soluble in alcohols | |
COC1COC2C(COC12)OC | |
174.20 | |
174.19 | |
Isosorbide dimethyl ether |
Safety and Handling
missing translation for 'einecsNumber' : 226-159-8
Product Identifier
- Isosorbide dimethyl ether
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title