missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 6200 H7
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Product Code | Brand | Voltage | Frequency | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Frequency | Price | Quantity & Availability | ||||
11306681
|
Fisherbrand™
211311100 |
230 V | 50 Hz |
N/A
|
N/A | ||||
Description
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters.
- High temperature and low level cut-outs
- Compact footprint maximizes bench space
Specifications
Force and Suction | |
10°C to 200°C |