missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ IsoTherm-System™
Used as an enzyme bar to prevent enzymes from becoming inactivated, as well as for PCR* sample preparation, sample transport and slow thawing of frozen samples.
Brand: Eppendorf™ 3880000160
This item is not returnable.
View return policy
Description
- Holds 24 tubes (0.5mL/1.5mL/2.0mL Safe-Lock microcentrifuge tubes or 1.7mL Safe-Twist screw cap tubes)
- IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
- Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
- No more melting ice baths (danger of contamination)
- All system components can be used separately
- Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor Specialises in cooling samples effectively and constantly at -21°C or 0°C over many hours.
Specifications
IsoPack and IsoRack set | |
0.5 mL | |
0°C | |
0.5mL tubes |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ IsoTherm-System™ > For use with: For 0.5mL tubes; Temperature Range (Freezer): 0deg.C
Spot an opportunity for improvement?Share a Content Correction