Learn More
Tuttnauer Isothermal/Disinfection (for Agar Preparation)
Product Code. 17809538 Shop All Tuttnauer Products
Click to view available options
:
Isothermal/Disinfection; Application: Agar Preparation
Product Code. 17809538
Brand: Tuttnauer ADD2220447
Only null left
Add to basket
Add to basket
Isothermal/Disinfection (for Agar preparation). The isothermal cycle has a temperature range from 60°C to 94°C and allows for gentle heating and cooling of agar.
  • Disinfection (agar prep)
  • Isothermal/Disinfection (for Agar preparation).
  • The isothermal cycle has a temperature range from 60°C to 94°C and allows for gentle heating and cooling of agar.
Specifications
For Use With (Application) Agar Preparation
Product Type Software