missing translation for 'onlineSavingsMsg'
Learn More

Isradipine, Tocris Bioscience™

Product Code. 18003802
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CaV1.x blocker

Potent and selective L-type voltage-gated Ca2+ channel blocker. EC50 and pA2 values are 1.4 nM and 10.3 for relaxation of depolarization- and Ca2+-induced contractions of rabbit aorta respectively; EC25 = 0.45 nM for reduction in rate of spontaneously beating guinea pig right atria. Long-acting antihypertensive agent in vivo, exerting primary effects on vascular tissue with secondary negative chronotropic action.

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5
Molecular Weight (g/mol) 371.39
MDL Number MFCD00153820
InChI Key HMJIYCCIJYRONP-UHFFFAOYNA-N
Synonym isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
PubChem CID 3784
IUPAC Name 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C

Specifications

Quantity 50 mg
Formula Weight 371.39
Percent Purity >99%
Chemical Name or Material Isradipine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.