Learn More
ITGA4 Monoclonal Antibody (R1-2), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA170075
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-70075 detects CD49d in mouse samples. MA1-70075 has been successfully used in flow cytometry, immunohistochemistry (frozen sections) and immunoprecipitation procedures. The MA1-70075 immunogen is Peyers Patch HEV binding lymphoma line (TK1).
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.Specifications
ITGA4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q00651 | |
ITGA4 | |
Peyers Patch HEV binding lymphoma line (TK1). | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
R1-2 | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Rat | |
Protein G | |
RUO | |
16401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |