missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPRIPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531770
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (90%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ITPRIPL1 Polyclonal antibody specifically detects ITPRIPL1 in Human samples. It is validated for Western Blot.Specifications
ITPRIPL1 | |
Polyclonal | |
Unconjugated | |
ITPRIPL1 | |
inositol 1,4,5-triphosphate receptor-interacting protein-like 1; inositol 1,4,5-trisphosphate receptor interacting protein-like 1; inositol 1,4,5-trisphosphate receptor-interacting protein-like 1; ITPRIPL1; KIAA1754L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6GPH6 | |
ITPRIPL1 | |
Recombinant fragment corresponding to a region within amino acids 311 and 555 of Human ITPRIPL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |