missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ J 113863
GREENER_CHOICE

Product Code. 30001443
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 30001443

Brand: Tocris Bioscience™ 2595/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent CCR1 chemokine receptor antagonist

Potent chemokine receptor 1 (CCR1) antagonist (IC50 values are 0.9 and 5.8 nM for human and mouse CCR1 receptors respectively). Also displays high selectivity for human but not mouse CCR3 receptors (IC50 values are 0.58 and 460 nM respectively). Improves paw inflammation, joint damage and dramatically reduces cell infiltration into joints in collagen-induced arthritis in mice.

Isomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353791-85-2
Molecular Formula C30H37Cl2IN2O2

Specifications

Quantity 1 mg
Formula Weight 655.44
Purity >98%
Molecular Weight (g/mol) 655.442
Chemical Name or Material J 113863
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.